siRNA Details
Gene Symbol: Slc30a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109088.1 | NP_001102558.1 | 14 | 1233 | ||
XM_006250963.1 | XP_006251025.1 | 14 | 1303 | |||
XM_006250965.2 | XP_006251027.1 | 14 | 1703 | |||
GenBank mRNA | FQ222791.1 | - | 12 | 1526 |
