siRNA Details
Gene Symbol: Ccdc117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017502.1 | NP_001017502.1 | 5 | 835 | ||
GenBank mRNA | BC087119.1 | - | 5 | 835 | 7097247 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017502.1 | NP_001017502.1 | 5 | 835 | ||
GenBank mRNA | BC087119.1 | - | 5 | 835 | 7097247 |