siRNA Details
Gene Symbol: Tns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170459.1 | NP_001163930.1 | 25, 26 | 4084 | ||
XM_006251459.1 | XP_006251521.1 | 27, 28 | 4470 | |||
XM_006251460.1 | XP_006251522.1 | 27, 28 | 4467 | |||
XM_006251462.2 | XP_006251524.1 | 27, 28 | 4852 | |||
XM_006251463.2 | XP_006251525.1 | 27, 28 | 4309 | |||
XM_006251464.2 | XP_006251526.1 | 27, 28 | 4401 | |||
XM_006251466.2 | XP_006251528.1 | 25, 26 | 4461 | |||
XM_006251467.2 | XP_006251529.1 | 27, 28 | 4698 | |||
XM_008770334.1 | XP_008768556.1 | 27, 28 | 5678 | |||
XM_008770335.1 | XP_008768557.1 | 28, 29 | 4870 | |||
XM_008770336.1 | XP_008768558.1 | 24, 25 | 5714 | |||
XM_008770337.1 | XP_008768559.1 | 25, 26 | 9556 | |||
XM_008770338.1 | XP_008768560.1 | 27, 28 | 4462 | |||
GenBank mRNA | AB510354.1 | - | 25, 26 | 4094 |
