siRNA Details
Gene Symbol: Tns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170459.1 | NP_001163930.1 | 29 | 4371 | ||
XM_006251459.1 | XP_006251521.1 | 31 | 4757 | |||
XM_006251460.1 | XP_006251522.1 | 31 | 4754 | |||
XM_006251462.2 | XP_006251524.1 | 31 | 5139 | |||
XM_006251463.2 | XP_006251525.1 | 31 | 4596 | |||
XM_006251464.2 | XP_006251526.1 | 31 | 4688 | |||
XM_006251466.2 | XP_006251528.1 | 29 | 4748 | |||
XM_006251467.2 | XP_006251529.1 | 31 | 4985 | |||
XM_008770334.1 | XP_008768556.1 | 31 | 5965 | |||
XM_008770335.1 | XP_008768557.1 | 32 | 5157 | |||
XM_008770336.1 | XP_008768558.1 | 28 | 6001 | |||
XM_008770337.1 | XP_008768559.1 | 29 | 9843 | |||
XM_008770338.1 | XP_008768560.1 | 31 | 4749 | |||
GenBank mRNA | AB510354.1 | - | 29 | 4381 | ||
FM131608.1 | - | 3 | 91 |
