siRNA Details
Gene Symbol: Gpr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109096.1 | NP_001102566.1 | 1 | 693 | ||
XM_006251616.2 | XP_006251678.1 | 1 | 798 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109096.1 | NP_001102566.1 | 1 | 693 | ||
XM_006251616.2 | XP_006251678.1 | 1 | 798 |