siRNA Details
Gene Symbol: Gnpnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134756.1 | NP_001128228.1 | 2 | 184 | ||
NM_001134757.1 | NP_001128229.1 | 1 | 190 | |||
XM_006251782.2 | XP_006251844.1 | 2 | 201 | |||
XM_006251783.2 | XP_006251845.1 | 2 | 249 | |||
XM_006251784.2 | XP_006251846.1 | 2 | 197 | |||
GenBank mRNA | BC160861.1 | - | 1 | 190 | 7322586 | |
CA339205.1 | - | 2 | 254 |
