siRNA Details
Gene Symbol: RGD1565222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109101.1 | NP_001102571.1 | 2 | 643 | ||
XM_006252004.2 | XP_006252066.1 | 2 | 941 | |||
XM_006252005.2 | XP_006252067.1 | 2 | 599 | |||
XM_006252006.2 | XP_006252068.1 | 559 |
