siRNA Details
Gene Symbol: RGD1561145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134605.2 | NP_001128077.2 | 3 | 199 | ||
XM_008771144.1 | XP_008769366.1 | 3 | 355 | |||
GenBank mRNA | CB315636.1 | - | 3 | 202 |
