siRNA Details
Gene Symbol: Slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109107.2 | NP_001102577.2 | 6 | 1063 | ||
GenBank mRNA | CB316326.1 | - | 4 | 569 | ||
EV776552.1 | - | 1 | 49 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109107.2 | NP_001102577.2 | 6 | 1063 | ||
GenBank mRNA | CB316326.1 | - | 4 | 569 | ||
EV776552.1 | - | 1 | 49 |