siRNA Details
Gene Symbol: Fam32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128078.1 | NP_001121550.1 | 1, 2 | 100 | ||
GenBank mRNA | BC166765.1 | - | 1, 2 | 100 | 7932317 | |
FQ219981.1 | - | 1, 2 | 122 | |||
FQ229768.1 | - | 1, 2 | 122 |
