siRNA Details
Gene Symbol: Kxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025143.1 | NP_001020314.1 | 2 | 199 | ||
XM_006252922.2 | XP_006252984.1 | 2 | 277 | |||
XM_006252923.2 | XP_006252985.1 | 2 | 604 | |||
XM_006252924.1 | XP_006252986.1 | 2 | 544 | |||
XM_006252925.1 | XP_006252987.1 | 141 | ||||
GenBank mRNA | BC088216.1 | - | 2 | 199 | 7383995 |
