siRNA Details
Gene Symbol: RGD1562024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113788.1 | NP_001107260.1 | 2 | 245 | ||
XM_006253581.2 | XP_006253643.1 | 2 | 1127 | |||
XM_006253582.2 | XP_006253644.1 | 2 | 2095 | |||
GenBank mRNA | BC158639.1 | - | 2 | 245 | 7457845 |
