siRNA Details
Gene Symbol: Fam193b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170408.1 | NP_001163879.1 | 7 | 1370 | ||
XM_006253646.2 | XP_006253708.1 | 6 | 1469 | |||
XM_006253647.2 | XP_006253709.1 | 6 | 2209 | |||
XM_006253650.2 | XP_006253712.1 | 6 | 2103 | |||
XM_006253651.2 | XP_006253713.1 | 7 | 2270 | |||
XM_008771503.1 | XP_008769725.1 | 6 | 3888 | |||
GenBank mRNA | CB717749.1 | - | 3 | 439 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145382, BC021381, AK173303, BC025483, AK155237, and others.
