siRNA Details
Gene Symbol: Fam193b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170408.1 | NP_001163879.1 | 8 | 1582 | ||
XM_006253646.2 | XP_006253708.1 | 7 | 1681 | |||
XM_006253647.2 | XP_006253709.1 | 7 | 2421 | |||
XM_006253650.2 | XP_006253712.1 | 7 | 2315 | |||
XM_006253651.2 | XP_006253713.1 | 8 | 2482 | |||
XM_008771503.1 | XP_008769725.1 | 7 | 4100 | |||
GenBank mRNA | BC166819.1 | - | 2 | 375 | 8367512 |
