siRNA Details
Gene Symbol: RGD1562987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173472.1 | NP_001166943.1 | 2, 3 | 282 | ||
XM_008772163.1 | XP_008770385.1 | 2, 3 | 260 | |||
GenBank mRNA | BC162045.1 | - | 2, 3 | 271 | 7107313 | |
CB568734.1 | - | 2, 3 | 283 | |||
FQ217314.1 | - | 2, 3 | 280 | |||
FQ224078.1 | - | 2, 3 | 270 | |||
FQ228113.1 | - | 2, 3 | 258 | |||
FQ229472.1 | - | 2, 3 | 240 |
