siRNA Details
Gene Symbol: Coq9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035257.1 | NP_001030334.1 | 5 | 555 | ||
GenBank mRNA | BC079370.1 | - | 5 | 541 | ||
BC104702.1 | - | 5 | 541 | |||
BC127449.1 | - | 5 | 556 | 8365327 | ||
CK600220.1 | - | 5 | 575 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026452, BC036386, AK009179, AK158979, AK154343.
Gene Ontology Categories:
