siRNA Details
Gene Symbol: Marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109132.1 | NP_001102602.1 | 2 | 501 | ||
XM_006255598.2 | XP_006255660.1 | 2 | 527 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109132.1 | NP_001102602.1 | 2 | 501 | ||
XM_006255598.2 | XP_006255660.1 | 2 | 527 |