siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227853.2 | XP_006227915.1 | 15 | 1945 | ||
XM_008758714.1 | XP_008756936.1 | 14 | 1931 | |||
XM_008758715.1 | XP_008756937.1 | 16 | 1998 | |||
XM_008758716.1 | XP_008756938.1 | 16 | 1996 | |||
GenBank mRNA | AY597251.1 | - | 2 | 341 | ||
X95466.1 | - | 1 | 253 |
