siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227853.2 | XP_006227915.1 | 18 | 2494 | ||
XM_008758714.1 | XP_008756936.1 | 17 | 2480 | |||
XM_008758715.1 | XP_008756937.1 | 19 | 2547 | |||
XM_008758716.1 | XP_008756938.1 | 19 | 2545 | |||
GenBank mRNA | AY597251.1 | - | 5 | 890 | ||
X95466.1 | - | 4 | 802 |
