siRNA Details
Gene Symbol: Smim17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109138.1 | NP_001102608.1 | 4 | 472 | ||
XM_006228194.2 | XP_006228256.1 | 4 | 511 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smim17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109138.1 | NP_001102608.1 | 4 | 472 | ||
XM_006228194.2 | XP_006228256.1 | 4 | 511 |