siRNA Details
Gene Symbol: LOC100909700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759061.1 | XP_008757283.1 | 3 | 297 |
Gene Symbol: Cd177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191948.1 | NP_001178877.1 | 3 | 257 | ||
XM_006228547.2 | XP_006228609.1 | 3 | 297 | |||
XM_008758969.1 | XP_008757191.1 | 3 | 297 |
