siRNA Details
Gene Symbol: Sertad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017513.1 | NP_001017513.1 | 2 | 91 | ||
XM_006228582.1 | XP_006228644.1 | 2 | 1065 | |||
GenBank mRNA | BC091229.1 | - | 2 | 91 | 7321753 |

More Information
Other Species Targeted

Gene Ontology Categories:
