siRNA Details
Gene Symbol: Igflr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134610.1 | NP_001128082.1 | 3 | 653 | ||
XM_006228786.2 | XP_006228848.1 | 3 | 862 | |||
XM_006228787.2 | XP_006228849.1 | 3 | 859 | |||
XM_006228788.2 | XP_006228850.1 | 3 | 577 |
