siRNA Details
Gene Symbol: LOC499235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574528.5 | XP_574528.1 | 1 | 39 | ||
GenBank mRNA | AY539892.1 | - | 1 | 39 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC499235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574528.5 | XP_574528.1 | 1 | 39 | ||
GenBank mRNA | AY539892.1 | - | 1 | 39 |