siRNA Details
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037793.2 | NP_001032882.2 | 8 | 969 | ||
XM_006231244.2 | XP_006231306.1 | 8 | 1180 | |||
XM_006231245.2 | XP_006231307.1 | 7 | 1744 | |||
XM_008760310.1 | XP_008758532.1 | 9 | 1235 | |||
XM_008760311.1 | XP_008758533.1 | 8 | 821 |
