siRNA Details
Gene Symbol: Plekhs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134613.1 | NP_001128085.1 | 5 | 308 | ||
XM_006231646.2 | XP_006231708.1 | 5 | 498 | |||
XM_006231647.2 | XP_006231709.1 | 5 | 498 | |||
XM_006231648.2 | XP_006231710.1 | 5 | 498 | |||
XM_008760543.1 | XP_008758765.1 | 5 | 495 | |||
XM_008760544.1 | XP_008758766.1 | 5 | 498 |
