siRNA Details
Gene Symbol: Clasrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024294.1 | NP_001019465.2 | 4 | 301 | ||
XM_008758970.1 | XP_008757192.1 | 4 | 372 | |||
XM_008758971.1 | XP_008757193.1 | 4 | 286 | |||
GenBank mRNA | BC089090.1 | - | 4 | 301 | 7111206 |
