siRNA Details
Gene Symbol: Hsf2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127683.1 | NP_001121155.1 | 2 | 139 | ||
XM_008772826.1 | XP_008771048.1 | 3 | 375 | |||
GenBank mRNA | BC158765.1 | - | 2 | 126 |
