siRNA Details
Gene Symbol: LOC103694873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773822.1 | XP_008772044.1 | 5 | 2105 | ||
XM_008773823.1 | XP_008772045.1 | 6 | 1785 | |||
XM_008773824.1 | XP_008772046.1 | 6 | 1782 | |||
XM_008773825.1 | XP_008772047.1 | 5 | 2102 | |||
XM_008773826.1 | XP_008772048.1 | 5 | 1618 | |||
XM_008773827.1 | XP_008772049.1 | 5 | 1615 |
Gene Symbol: RGD1564162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256332.2 | XP_006256394.1 | 6 | 1785 | ||
XM_006256333.2 | XP_006256395.1 | 6 | 1782 | |||
XM_006256335.2 | XP_006256397.1 | 5 | 1618 | |||
XM_006256336.2 | XP_006256398.1 | 5 | 1615 | |||
XM_008772934.1 | XP_008771156.1 | 5 | 2105 | |||
XM_008772935.1 | XP_008771157.1 | 5 | 2102 | |||
GenBank mRNA | BC160832.1 | - | 6 | 1049 |
