siRNA Details
Gene Symbol: Lrrc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109171.1 | NP_001102641.1 | 2 | 207 | ||
XM_006256465.2 | XP_006256527.1 | 2 | 230 | |||
XM_006256466.2 | XP_006256528.1 | 2 | 1022 | |||
XM_006256467.2 | XP_006256529.1 | 2 | 251 | |||
XM_006256468.2 | XP_006256530.1 | 2 | 263 | |||
GenBank mRNA | FQ223562.1 | - | 2 | 201 |
