siRNA Details
Gene Symbol: Lrrc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109171.1 | NP_001102641.1 | 5 | 592 | ||
XM_006256465.2 | XP_006256527.1 | 5 | 618 | |||
XM_006256466.2 | XP_006256528.1 | 5 | 1410 | |||
XM_006256467.2 | XP_006256529.1 | 5 | 639 | |||
XM_006256468.2 | XP_006256530.1 | 5 | 651 | |||
XM_008772902.1 | XP_008771124.1 | 4 | 414 | |||
XM_008772903.1 | XP_008771125.1 | 4 | 1381 | |||
GenBank mRNA | FQ223562.1 | - | 5 | 586 |
