siRNA Details
Gene Symbol: LOC499469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047940.1 | NP_001041405.1 | 4 | 429 | ||
XM_008773005.1 | XP_008771227.1 | 2 | 359 | |||
GenBank mRNA | AY325181.1 | - | 4 | 429 |
