siRNA Details
Gene Symbol: Ndufs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025146.1 | NP_001020317.1 | 4 | 407 | ||
GenBank mRNA | BC083729.1 | - | 4 | 407 | 7191329 | |
BC127458.1 | - | 4 | 407 | 7193742 | ||
FQ212751.1 | - | 4 | 424 | |||
FQ214408.1 | - | 4 | 424 | |||
FQ214557.1 | - | 4 | 424 | |||
FQ216263.1 | - | 4 | 407 | |||
FQ223158.1 | - | 4 | 424 | |||
FQ223379.1 | - | 4 | 424 |
