siRNA Details
Gene Symbol: LOC499530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047941.1 | NP_001041406.1 | 3, 4 | 299 | ||
XM_008760739.1 | XP_008758961.1 | 3, 4 | 299 | |||
GenBank mRNA | AY539940.1 | - | 3, 4 | 299 |
