siRNA Details
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109176.1 | NP_001102646.1 | 2, 3 | 1359 | ||
XM_006232019.2 | XP_006232081.1 | 3, 4 | 1639 | |||
XM_008760784.1 | XP_008759006.1 | 3, 4 | 1640 | |||
XM_008760785.1 | XP_008759007.1 | 3, 4 | 1641 | |||
XM_008760786.1 | XP_008759008.1 | 4, 5 | 1733 | |||
XM_008760787.1 | XP_008759009.1 | 3, 4 | 3093 | |||
XM_008760788.1 | XP_008759010.1 | 3, 4 | 3188 | |||
XM_008760789.1 | XP_008759011.1 | 3, 4 | 1639 | |||
XM_008760790.1 | XP_008759012.1 | 3, 4 | 1641 | |||
GenBank mRNA | FQ232832.1 | - | 2, 3 | 1157 |
