siRNA Details
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109176.1 | NP_001102646.1 | 1 | 258 | ||
XM_006232019.2 | XP_006232081.1 | 2 | 538 | |||
XM_008760784.1 | XP_008759006.1 | 2 | 539 | |||
XM_008760785.1 | XP_008759007.1 | 2 | 540 | |||
XM_008760786.1 | XP_008759008.1 | 3 | 632 | |||
XM_008760787.1 | XP_008759009.1 | 2 | 1992 | |||
XM_008760788.1 | XP_008759010.1 | 2 | 2087 | |||
XM_008760789.1 | XP_008759011.1 | 2 | 538 | |||
XM_008760790.1 | XP_008759012.1 | 2 | 540 | |||
GenBank mRNA | FQ232832.1 | - | 1 | 56 |
