siRNA Details
Gene Symbol: LOC499565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047946.1 | NP_001041411.1 | 4 | 358 | ||
GenBank mRNA | AY383693.1 | - | 4 | 358 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC499565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047946.1 | NP_001041411.1 | 4 | 358 | ||
GenBank mRNA | AY383693.1 | - | 4 | 358 |