siRNA Details
Gene Symbol: Lekr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115029.2 | NP_001108501.2 | 13 | 2320 | ||
XM_006232472.2 | XP_006232534.1 | 12 | 3135 | |||
XM_006232473.1 | XP_006232535.1 | 13 | 1982 | |||
XM_006232474.2 | XP_006232536.1 | 12 | 2194 | |||
XM_006232475.2 | XP_006232537.1 | 12 | 1845 |
