siRNA Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109190.2 | NP_001102660.1 | 4 | 1109 | ||
XM_006232897.2 | XP_006232959.1 | 4 | 1035 | |||
XM_006232898.2 | XP_006232960.1 | 4 | 1033 | |||
XM_006232900.2 | XP_006232962.1 | 4 | 1032 | |||
XM_006232902.2 | XP_006232964.1 | 4 | 1035 | |||
XM_006232903.2 | XP_006232965.1 | 3 | 1406 | |||
XM_006232904.2 | XP_006232966.1 | 5 | 2763 | |||
XM_006232905.2 | XP_006232967.1 | 5 | 2796 | |||
XM_006232906.2 | XP_006232968.1 | 3 | 3868 | |||
XM_006232907.2 | XP_006232969.1 | 3 | 353 | |||
XM_006232908.2 | XP_006232970.1 | 4 | 1033 | |||
XM_006232911.2 | XP_006232973.1 | 3 | 230 | |||
XM_006232912.2 | XP_006232974.1 | 2 | 152 | |||
XM_008761322.1 | XP_008759544.1 | 5 | 3350 | |||
XM_008761323.1 | XP_008759545.1 | 4 | 319 | |||
XM_008761324.1 | XP_008759546.1 | 3 | 2473 | |||
XM_008761325.1 | XP_008759547.1 | 3 | 2427 | |||
XM_008761326.1 | XP_008759548.1 | 3 | 2424 |
