siRNA Details
Gene Symbol: Ccdc183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025040.2 | NP_001020211.2 | 10, 11 | 1137 | ||
XM_006233635.1 | XP_006233697.1 | 11, 12 | 1786 | |||
GenBank mRNA | BC160897.1 | - | 10, 11 | 1137 | 7453119 |
