siRNA Details
Gene Symbol: LOC499886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024312.1 | NP_001019483.1 | 6 | 585 | ||
XM_008762191.1 | XP_008760413.1 | 6 | 641 | |||
XM_008762192.1 | XP_008760414.1 | 6 | 641 | |||
GenBank mRNA | BC086511.1 | - | 6 | 585 | 7132989 |
