siRNA Details
Gene Symbol: Ap5s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134618.1 | NP_001128090.1 | 3 | 543 | ||
XM_006235038.2 | XP_006235100.1 | 3 | 562 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ap5s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134618.1 | NP_001128090.1 | 3 | 543 | ||
XM_006235038.2 | XP_006235100.1 | 3 | 562 |