siRNA Details
Gene Symbol: Tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134619.1 | NP_001128091.1 | 1 | 1886 | ||
XM_006235428.2 | XP_006235490.1 | 2 | 1963 | |||
XM_008762375.1 | XP_008760597.1 | 2 | 2726 |
