siRNA Details
Gene Symbol: RGD1561282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109213.1 | NP_001102683.1 | 2 | 290 | ||
XM_008762534.1 | XP_008760756.1 | 3 | 2751 | |||
XM_008762535.1 | XP_008760757.1 | 3 | 2751 | |||
XM_008762536.1 | XP_008760758.1 | 3 | 1472 | |||
XM_008762537.1 | XP_008760759.1 | 3 | 2751 | |||
XM_008762538.1 | XP_008760760.1 | 3 | 2751 | |||
XM_008762539.1 | XP_008760761.1 | 3 | 1472 |
