siRNA Details
Gene Symbol: Asb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109216.2 | NP_001102686.1 | 1 | 372 | ||
GenBank mRNA | BC168867.1 | - | 1 | 372 | 7191440 |

More Information
Other Species Targeted

Gene Ontology Categories:
