siRNA Details
Gene Symbol: Cldn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100813.1 | NP_001094283.1 | 2 | 431 | ||
XM_006236052.2 | XP_006236114.1 | 4 | 656 | |||
XM_006236053.2 | XP_006236115.1 | 4 | 638 |
