siRNA Details
Gene Symbol: RGD1563091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236079.2 | XP_006236141.1 | 2 | 3978 | ||
XM_575365.5 | XP_575365.2 | 3 | 4243 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1563091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236079.2 | XP_006236141.1 | 2 | 3978 | ||
XM_575365.5 | XP_575365.2 | 3 | 4243 |