siRNA Details
Gene Symbol: Phf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110492.1 | NP_001103962.1 | 11, 12 | 2464 | ||
XM_006236104.2 | XP_006236166.1 | 11, 12 | 2500 | |||
XM_006236105.2 | XP_006236167.1 | 11, 12 | 2239 | |||
XM_006236107.2 | XP_006236169.1 | 11, 12 | 2500 | |||
XM_006236108.2 | XP_006236170.1 | 11, 12 | 2500 | |||
XM_006236109.2 | XP_006236171.1 | 11, 12 | 2500 | |||
XM_006236111.2 | XP_006236173.1 | 11, 12 | 2499 | |||
XM_008762716.1 | XP_008760938.1 | 11, 12 | 2500 | |||
GenBank mRNA | BC166456.1 | - | 11, 12 | 2195 | 9025134 | |
FQ209867.1 | - | 11, 12 | 2428 | |||
FQ221829.1 | - | 10, 11 | 2742 |
