siRNA Details
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236277.2 | XP_006236339.1 | 17, 18 | 4000 | ||
XM_006236278.2 | XP_006236340.1 | 16, 17 | 5589 | |||
XM_008762844.1 | XP_008761066.1 | 16, 17 | 5591 |
