siRNA Details
Gene Symbol: Plekha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109235.2 | NP_001102705.1 | 5 | 678 | ||
XM_006236506.2 | XP_006236568.1 | 5 | 763 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plekha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109235.2 | NP_001102705.1 | 5 | 678 | ||
XM_006236506.2 | XP_006236568.1 | 5 | 763 |